(E/Z)-4-Hydroxytamoxifen 98%

Supplier: Ambeed
Danger

Synonyms: Afimoxifene, 4-OHT, 4-[1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]-phenol, 4-(1-(4-(2-(Dimethylamino)ethoxy)phenyl)-2-phenylbut-1-en-1-yl)phenol

A982086-1G A982086-250MG A982086-100MG
76656-664EA 381 USD
76656-664 76656-666 77784-491
(E/Z)-4-Hydroxytamoxifen 98%
(E/Z)-4-Hydroxytamoxifen

Precaución: for Research Use Only

Formula: C₂₆H₂₉NO₂
MDL Number: MFCD00278780
CAS Number: 68392-35-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR