3-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-5-methyl-6-(pyridin-3-yl)-3H-imidazo[4,5-b]pyridine 98%

Supplier: Ambeed
Danger

Synonyms: TM-25659

A981573-10MG A981573-1MG A981573-25MG A981573-5MG
76656-654EA 50 USD
76656-654 76656-656 76656-658 76656-660
3-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-5-methyl-6-(pyridin-3-yl)-3H-imidazo[4,5-b]pyridine 98%
3-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-5-methyl-6-(pyridin-3-yl)-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₃₀H₂₈N₈
MW: 500.6 g/mol
MDL Number: MFCD32671933
CAS Number: 260553-97-7
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR