(R)-2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)butoxy)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: RGX-104

A981571-100MG A981571-10MG A981571-1MG A981571-25MG A981571-50MG A981571-5MG
76656-642EA 288.57 USD
76656-642 76656-644 76656-646 76656-648 76656-650 76656-652
(R)-2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)butoxy)phenyl)acetic acid 98%
(R)-2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)butoxy)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₃₄H₃₃ClF₃NO₃
MW: 596.08 g/mol
MDL Number: MFCD31726291
CAS Number: 610318-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR