(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 99+%

Supplier: Ambeed
Warning

Synonyms: Rasagiline, (1R)-N-prop-2-ynylindan-1-amine

A981553-100MG A981553-1MG A981553-50MG
76656-636EA 13 USD
76656-636 76656-638 76656-640
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 99+%
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.2383 g/mol
MDL Number: MFCD00866571
CAS Number: 136236-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR