1-(6-Cyclopropylpyridin-3-yl)ethanamine 97%

Supplier: Ambeed
Warning

A981535-1G A981535-250MG
76656-626EA 347.98 USD
76656-626 76656-628
1-(6-Cyclopropylpyridin-3-yl)ethanamine 97%
1-(6-Cyclopropylpyridin-3-yl)ethanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MW: 162.2316 g/mol
CAS Number: 1256821-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR