2-Ethyl-6-methoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: Naproxen EP Impurity J

A981498-1G A981498-250MG
76656-622EA 125.19 USD
76656-622 76656-624
2-Ethyl-6-methoxynaphthalene 98%
2-Ethyl-6-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₃H₁₄O
MW: 186.2497 g/mol
MDL Number: MFCD08704412
CAS Number: 21388-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR