SPHINX31 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(4-(Pyridin-2-ylmethyl)piperazin-1-yl)-5-(trifluoromethyl)phenyl)-5-(pyridin-4-yl)furan-2-carboxamide

A981488-10MG A981488-1G A981488-1MG A981488-250MG A981488-25MG A981488-50MG A981488-5MG
76656-600EA 136.86 USD
76656-600 76656-602 76656-604 76656-606 76656-608 76656-610 76656-612
SPHINX31 98%
SPHINX31

Precaución: for Research Use Only

Formula: C₂₇H₂₄F₃N₅O₂
MW: 507.52 g/mol
MDL Number: MFCD31810499
CAS Number: 1818389-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR