Tenatoprazole sodium salt 98%

Supplier: Ambeed
Warning

A981468-10MG A981468-1MG A981468-25MG A981468-5MG
76656-592EA 23 USD
76656-592 76656-594 76656-596 76656-598
Tenatoprazole sodium salt 98%
Tenatoprazole sodium salt

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄NaO₃S
MDL Number: MFCD08059005
CAS Number: 335299-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR