1,3,5-Tri(pyridin-4-yl)benzene 97%

Supplier: Ambeed
Warning

A981457-100MG A981457-1G A981457-250MG A981457-5G
76656-582EA 19.1 USD
76656-582 76656-584 76656-586 76656-588
1,3,5-Tri(pyridin-4-yl)benzene 97%
1,3,5-Tri(pyridin-4-yl)benzene

Precaución: for Research Use Only

Formula: C₂₁H₁₅N₃
MW: 309.3639 g/mol
MDL Number: MFCD29049593
CAS Number: 170165-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR