3,5-Di(pyridin-4-yl)-4H-1,2,4-triazol-4-amine 97%

Supplier: Ambeed
Warning

A981446-100MG A981446-1G A981446-250MG
76656-576EA 49.87 USD
76656-576 76656-578 76656-580
3,5-Di(pyridin-4-yl)-4H-1,2,4-triazol-4-amine 97%
3,5-Di(pyridin-4-yl)-4H-1,2,4-triazol-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₆
MW: 238.2480 g/mol
MDL Number: MFCD03017904
CAS Number: 38634-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR