5,5-Dimethyl-1,3,2-dioxathiane 2,2-dioxide 97%

Supplier: Ambeed
Danger

A1350488-1G A1350488-250MG
76658-186EA 47.75 USD
76658-186 76658-188
5,5-Dimethyl-1,3,2-dioxathiane 2,2-dioxide 97%
5,5-Dimethyl-1,3,2-dioxathiane 2,2-dioxide

Precaución: for Research Use Only

Formula: C₅H₁₀O₄S
MW: 166.1955 g/mol
MDL Number: MFCD02661105
CAS Number: 1755-97-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR