Tris(dibenzylideneacetone)dipalladium(0) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]
A980151-1G
A980151-250MG
A980151-5G
76656-546EA
63.66
USD
76656-546
76656-548
76656-550
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)
Precaución: for Research Use Only
Formula:
C₅₁H₄₂O₃Pd₂ MW: 915.72 g/mol |
MDL Number:
MFCD00013310 CAS Number: 51364-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...