4-Bromo-trans-stilbene 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: trans-4-Bromostilbene

A979993-1G A979993-250MG A979993-5G
76656-534EA 101.85 USD
76656-534 76656-536 76656-538
4-Bromo-trans-stilbene 98% (by GC)
4-Bromo-trans-stilbene

Precaución: for Research Use Only

Formula: C₁₄H₁₁Br
MW: 259.15 g/mol
MDL Number: MFCD00155020
CAS Number: 13041-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR