2-(tert-Butoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A979484-1G A979484-250MG A979484-5G
76656-498EA 161.26 USD
76656-498 76656-500 76656-502
2-(tert-Butoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%
2-(tert-Butoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₃
MW: 277.1670 g/mol
MDL Number: MFCD11878215
CAS Number: 1421341-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR