1-[(tert-Butoxy)carbonyl]-3-ethylpyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A978814-100MG A978814-1G A978814-250MG
76656-490EA 63.66 USD
76656-490 76656-492 76656-494
1-[(tert-Butoxy)carbonyl]-3-ethylpyrrolidine-3-carboxylic acid 97%
1-[(tert-Butoxy)carbonyl]-3-ethylpyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
MDL Number: MFCD12406578
CAS Number: 1158751-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR