[1,3]Dioxolo[4,5-b]pyridin-6-ylboronic acid 98%

Supplier: Ambeed
Warning

A978738-100MG A978738-250MG
76656-486EA 175.05 USD
76656-486 76656-488
[1,3]Dioxolo[4,5-b]pyridin-6-ylboronic acid 98%
[1,3]Dioxolo[4,5-b]pyridin-6-ylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BNO₄
MW: 166.93 g/mol
MDL Number: MFCD18250790
CAS Number: 1019767-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR