PKM2-IN-1 98%

Supplier: Ambeed
Warning

Synonyms: MDK4882, PKM2 inhibitor, (3-Methyl-1,4-dioxo-1,4-dihydronaphthalen-2-yl)methyl piperidine-1-carbodithioate, (3-methyl-1,4-dioxonaphthalen-2-yl)methyl piperidine-1-carbodithioate, Compound 3k

A976997-10MG A976997-1MG A976997-25MG A976997-50MG A976997-5MG A976997-250MG A976997-1G A976997-100MG
76656-462EA 27 USD
76656-462 76656-464 76656-466 76656-468 76656-470 77623-232 77784-462 77784-461
PKM2-IN-1 98%
PKM2-IN-1

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₂S₂
MW: 345.47 g/mol
MDL Number: MFCD31619244
CAS Number: 94164-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR