1-Butyl-3-methylimidazolium thiocyanate 99%

Supplier: Ambeed
Warning

A976228-1G A976228-5G A976228-25G
76656-450EA 6 USD
76656-450 76656-452 77784-459
1-Butyl-3-methylimidazolium thiocyanate 99%
1-Butyl-3-methylimidazolium thiocyanate

Precaución: for Research Use Only

Formula: C₉H₁₅N₃S
MDL Number: MFCD06798181
CAS Number: 344790-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR