Propylamine hydrobromide 97% (by total nitrogen and titration analysis)

Supplier: Ambeed
Warning

A975468-1G A975468-25G A975468-5G
76656-444EA 7 USD
76656-444 76656-446 76656-448
Propylamine hydrobromide 97% (by total nitrogen and titration analysis)
Propylamine hydrobromide

Precaución: for Research Use Only

Formula: C₃H₉N·HBr
MDL Number: MFCD30749306
CAS Number: 4905-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR