Dimethyldiphenylsilane 97% (by GC)

Supplier: Ambeed
Warning

A975180-1G A975180-250MG A975180-5G
76656-438EA 14 USD
76656-438 76656-440 76656-442
Dimethyldiphenylsilane 97% (by GC)
Dimethyldiphenylsilane

Precaución: for Research Use Only

Formula: C₁₄H₁₆Si
MDL Number: MFCD00041326
CAS Number: 778-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR