(2,2'-Bipyridyl)bis[2-(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A974809-100MG A974809-1G A974809-250MG
76656-428EA 56 USD
76656-428 76656-430 76656-432
(2,2'-Bipyridyl)bis[2-(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate 97%
(2,2'-Bipyridyl)bis[2-(4-fluorophenyl)pyridine]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₂H₂₂F₈IrN₄P
MW: 837.7200 g/mol
CAS Number: 1580547-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR