Lithium perfluorobutane-1-sulfonate 98% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: Lithium Nonafluoro-1-butanesulphonate, Lithium nonafluorobutanesulphonate

A974039-25G A974039-100G A974039-5G
76656-426EA 58.35 USD
76656-426 77599-330 77623-228
Lithium perfluorobutane-1-sulfonate 98% (by titrimetric analysis)
Lithium perfluorobutane-1-sulphonate

Precaución: for Research Use Only

Formula: C₄F₉LiO₃S
MW: 306.0326 g/mol
MDL Number: MFCD03094160
CAS Number: 131651-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR