N-(2,6-Dichloropyridin-4-yl)-2-(4-isopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-6-yl)hydrazine-1-carboxamide 98%
Supplier: Ambeed
|
Danger
|
A973785-10MG
A973785-1MG
A973785-25MG
76656-416EA
115
USD
76656-416
76656-418
76656-420
N-(2,6-Dichloropyridin-4-yl)-2-(4-isopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-6-yl)hydrazine-1-carboxamide 98%
N-(2,6-Dichloropyridin-4-yl)-2-(4-isopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-6-yl)hydrazine-1-carboxamide
Precaución: for Research Use Only
Formula:
C₁₇H₁₉Cl₂N₇O |
MDL Number:
MFCD09878275 CAS Number: 383150-41-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...