5-Acetyl-1-(4-methoxyphenyl)-6-methyl-3,4-dihydro-2(1H)-pyridinone 90%

Supplier: Ambeed
Warning

A972378-100MG A972378-250MG A972378-50MG
76656-410EA 705.5 USD
76656-410 76656-412 76656-414
5-Acetyl-1-(4-methoxyphenyl)-6-methyl-3,4-dihydro-2(1H)-pyridinone 90%
5-Acetyl-1-(4-methoxyphenyl)-6-methyl-3,4-dihydro-2(1H)-pyridinone

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₃
MW: 259.3004 g/mol
CAS Number: 171506-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR