6-(Chloromethyl)benzothiazole 95%

Supplier: Ambeed
Danger

Synonyms: 6-(Chloromethyl)-1,3-benzothiazole, Benzothiazole, 6-(chloromethyl)-, 6-(Chloromethyl)benzo[d]thiazole

A971466-1G A971466-250MG A971466-100MG
76656-402EA 206 USD
76656-402 77749-808 77749-807
6-(Chloromethyl)benzothiazole 95%
6-(Chloromethyl)benzothiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MDL Number: MFCD12756627
CAS Number: 149440-49-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR