3-(4-Hydroxy-3-(5,5,8,8-tetramethyl-3-(pentyloxy)-5,6,7,8-tetrahydronaphthalen-2-yl)phenyl)acrylic acid 98%

Supplier: Ambeed
Warning

Synonyms: UVI 3003

A971133-10MG A971133-1MG A971133-25MG A971133-50MG A971133-5MG
76656-390EA 110.01 USD
76656-390 76656-392 76656-394 76656-396 76656-398
3-(4-Hydroxy-3-(5,5,8,8-tetramethyl-3-(pentyloxy)-5,6,7,8-tetrahydronaphthalen-2-yl)phenyl)acrylic acid 98%
3-(4-Hydroxy-3-(5,5,8,8-tetramethyl-3-(pentyloxy)-5,6,7,8-tetrahydronaphthalen-2-yl)phenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₆O₄
MDL Number: MFCD18086862
CAS Number: 847239-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR