Bis(1-methyl-1H-imidazol-2-yl)methane 97%

Supplier: Ambeed
Warning

A1350486-100MG A1350486-1G A1350486-250MG A1350486-50MG
76658-178EA 225.98 USD
76658-178 76658-180 76658-182 76658-184
Bis(1-methyl-1H-imidazol-2-yl)methane 97%
Bis(1-methyl-1H-imidazol-2-yl)methane

Precaución: for Research Use Only

Formula: C₉H₁₂N₄
MW: 176.2184 g/mol
MDL Number: MFCD08668067
CAS Number: 124225-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR