Methyl 2-bromo-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A969849-1G A969849-250MG A969849-5G
76656-372EA 161.26 USD
76656-372 76656-374 76656-376
Methyl 2-bromo-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%
Methyl 2-bromo-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BBrO₄
MW: 355.0319 g/mol
MDL Number: MFCD19237219
CAS Number: 2096341-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR