Di-o-tolylamine 97%

Supplier: Ambeed
Warning

A968819-100MG A968819-1G A968819-250MG
76656-354EA 45.62 USD
76656-354 76656-356 76656-358
Di-o-tolylamine 97%
Di-o-tolylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
MW: 197.2756 g/mol
MDL Number: MFCD01851353
CAS Number: 617-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR