3-[2-(Benzyloxy)ethoxy]propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl-PEG2-acid

A967977-250MG A967977-100MG A967977-1G A967977-5G
76656-336EA 44.56 USD
76656-336 77618-658 77618-660 77749-804
3-[2-(Benzyloxy)ethoxy]propanoic acid 98%
3-[2-(Benzyloxy)ethoxy]propanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.2530 g/mol
MDL Number: MFCD28096691
CAS Number: 91555-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR