Imidazo[1,2-a]pyridin-8-ylmethanamine 95%

Supplier: Ambeed
Warning

A967012-250MG A967012-100MG A967012-1G
76656-326EA 293.87 USD
76656-326 77749-801 77749-802
Imidazo[1,2-a]pyridin-8-ylmethanamine 95%
Imidazo[1,2-a]pyridin-8-ylmethanamine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.18 g/mol
MDL Number: MFCD10698077
CAS Number: 933721-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR