NS 1738 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, α7 nAChR positive allosteric modulator

A966598-100MG A966598-10MG A966598-1MG A966598-25MG A966598-50MG A966598-5MG
76656-314EA 169 USD
76656-314 76656-316 76656-318 76656-320 76656-322 76656-324
NS 1738 98%
NS 1738

Precaución: for Research Use Only

Formula: C₁₄H₉Cl₂F₃N₂O₂
MW: 365.1 g/mol
MDL Number: MFCD11100189
CAS Number: 501684-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR