[(5-Bromopentyl)oxy](tert-butyl)dimethylsilane 95%

Supplier: Ambeed
Danger

A966315-1G A966315-250MG A966315-5G
76656-308EA 165.51 USD
76656-308 76656-310 76656-312
[(5-Bromopentyl)oxy](tert-butyl)dimethylsilane 95%
[(5-Bromopentyl)oxy](tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₂₅BrOSi
MW: 281.3051 g/mol
MDL Number: MFCD20261073
CAS Number: 85514-43-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR