[(1-Methyl-1H-benzimidazol-2-yl)methyl]amine dihydrochloride 95%

Supplier: Ambeed
Warning

A965446-100MG A965446-1G A965446-250MG A965446-5G
76656-286EA 36.08 USD
76656-286 76656-288 76656-290 76656-292
[(1-Methyl-1H-benzimidazol-2-yl)methyl]amine dihydrochloride 95%
[(1-Methyl-1H-benzimidazol-2-yl)methyl]amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃Cl₂N₃
MW: 234.1256 g/mol
MDL Number: MFCD06687785
CAS Number: 53332-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR