1-(3-Bromopropyl)-1H-imidazole hydrobromide 95%

Supplier: Ambeed
Danger

A965321-100MG A965321-1G A965321-250MG
76656-280EA 28 USD
76656-280 76656-282 76656-284
1-(3-Bromopropyl)-1H-imidazole hydrobromide 95%
1-(3-Bromopropyl)-1H-imidazole hydrobromide

Precaución: for Research Use Only

Formula: C₆H₁₀Br₂N₂
MDL Number: MFCD18917307
CAS Number: 144385-79-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR