2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid 97%

Supplier: Ambeed
Warning

A965126-1G A965126-250MG A965126-5G
76656-274EA 216 USD
76656-274 76656-276 76656-278
2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid 97%
2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BN₂O₄
MDL Number: MFCD13188969
CAS Number: 903513-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR