S-Methyl-S-(2-pyridinyl) sulfoximine 98%

Supplier: Ambeed
Warning

A965120-100MG A965120-250MG
76656-270EA 74.27 USD
76656-270 76656-272
S-Methyl-S-(2-pyridinyl) sulfoximine 98%
S-Methyl-S-(2-pyridinyl) sulfoximine

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MW: 156.21 g/mol
MDL Number: MFCD09878530
CAS Number: 76456-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR