9-Azabicyclo[3.3.1]nonan-3-one N-oxyl 98%

Supplier: Ambeed
Warning

A965115-1G A965115-250MG A965115-5G
76656-264EA 189 USD
76656-264 76656-266 76656-268
9-Azabicyclo[3.3.1]nonan-3-one N-oxyl 98%
9-Azabicyclo[3.3.1]nonan-3-one N-oxyl

Precaución: for Research Use Only

Formula: C₈H₁₂NO₂
MDL Number: MFCD29059815
CAS Number: 7123-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR