tert-Butyl (R)-4,5-diamino-5-oxopentanoate monohydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Glu(OtBu)-NH₂·HCl

A965113-25G A965113-5G
76656-260EA 340.55 USD
76656-260 76656-262
tert-Butyl (R)-4,5-diamino-5-oxopentanoate monohydrochloride 97%
tert-Butyl (R)-4,5-diamino-5-oxopentanoate monohydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₃
MW: 238.7118 g/mol
MDL Number: MFCD30491706
CAS Number: 66575-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR