Tetradecyl methacrylate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A965101-100G A965101-25G A965101-500G
76656-258EA 19 USD
76656-258 77784-440 77784-441
Tetradecyl methacrylate 98% stabilized
Tetradecyl methacrylate

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₂
MDL Number: MFCD00085280
CAS Number: 2549-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR