4-((5,5-Dimethyl-8-(p-tolyl)-5,6-dihydronaphthalen-2-yl)ethynyl)benzoic acid ≥98%

Supplier: Ambeed
Warning

A964784-10MG A964784-1MG A964784-25MG A964784-5MG
76656-240EA 165 USD
76656-240 76656-242 76656-244 76656-246
4-((5,5-Dimethyl-8-(p-tolyl)-5,6-dihydronaphthalen-2-yl)ethynyl)benzoic acid ≥98%
4-((5,5-Dimethyl-8-(p-tolyl)-5,6-dihydronaphthalen-2-yl)ethynyl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₄O₂
MDL Number: MFCD00943817
CAS Number: 171746-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR