m-Toluic acid 97%, (Chunk or granule or flakes)
Supplier: Ambeed
|
Warning
|
A964007-1000G
A964007-500G
A964007-100G
76656-224EA
17
USD
76656-224
76656-226
77749-797
m-Toluic acid 97%, (Chunk or granule or flakes)
m-Toluic acid
Precaución: for Research Use Only
Formula:
C₈H₈O₂ |
MDL Number:
MFCD00002523 CAS Number: 99-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...