2-((3-Methoxyphenyl)ethynyl)-6-methylpyridine hydrochloride 98%

Supplier: Ambeed
Warning

A963537-10MG A963537-1MG A963537-5MG
76656-218EA 17 USD
76656-218 76656-220 76656-222
2-((3-Methoxyphenyl)ethynyl)-6-methylpyridine hydrochloride 98%
2-((3-Methoxyphenyl)ethynyl)-6-methylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₄ClNO
MDL Number: MFCD19440696
CAS Number: 823198-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR