(1-Bromo-2,2,2-trifluoroethyl)benzene 95%

Supplier: Ambeed
Warning

A963368-100MG A963368-1G A963368-250MG
76656-212EA 47 USD
76656-212 76656-214 76656-216
(1-Bromo-2,2,2-trifluoroethyl)benzene 95%
(1-Bromo-2,2,2-trifluoroethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MDL Number: MFCD12789673
CAS Number: 434-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR