(3-Bromo-2-methyl-phenyl)-carbamic acid tert-butyl ester 97%
Supplier: Ambeed
|
Warning
|
A963143-100MG
A963143-1G
A963143-250MG
A963143-5G
76656-204EA
43.5
USD
76656-204
76656-206
76656-208
76656-210
(3-Bromo-2-methyl-phenyl)-carbamic acid tert-butyl ester 97%
(3-Bromo-2-methyl-phenyl)-carbamic acid tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BrNO₂ MW: 286.1649 g/mol |
MDL Number:
MFCD11975627 CAS Number: 221538-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...