6-((2-Amino-4-hydroxy-5-methylphenyl)diazenyl)-3,4-dihydroquinolin-2(1H)-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E/Z)-ZL0420
A962978-10MG
A962978-1MG
A962978-25MG
A962978-5MG
76656-196EA
56.23
USD
76656-196
76656-198
76656-200
76656-202
6-((2-Amino-4-hydroxy-5-methylphenyl)diazenyl)-3,4-dihydroquinolin-2(1H)-one 98%
6-((2-Amino-4-hydroxy-5-methylphenyl)diazenyl)-3,4-dihydroquinolin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₁₆H₁₆N₄O₂ MW: 296.32 g/mol |
MDL Number:
MFCD31807601 CAS Number: 2229039-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...