8-Aminooctanoic acid tert-butyl ester 95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 8-aminooctanoate

A962583-250MG A962583-1G A962583-100MG
76656-182EA 178.24 USD
76656-182 77573-904 77623-200
8-Aminooctanoic acid tert-butyl ester 95%
8-Aminooctanoic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₅NO₂
MW: 215.3324 g/mol
MDL Number: MFCD24624132
CAS Number: 102522-32-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR