2,5,6-Trichlorobenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A961606-100MG A961606-250MG
76656-170EA 276.9 USD
76656-170 77749-793
2,5,6-Trichlorobenzo[d]thiazole 95%
2,5,6-Trichlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₂Cl₃NS
MW: 238.5215 g/mol
MDL Number: MFCD09749269
CAS Number: 120258-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR