2-(2-Propynyloxy)ethylamine 98% (by titrimetric analysis)
Supplier: Ambeed
|
Danger
|
Synonyms:
Propargyl-PEG1-amine, Propargyl-PEG1-NH2
A961334-100MG
A961334-1G
A961334-250MG
A961334-5G
A961334-25G
76656-162EA
16
USD
76656-162
76656-164
76656-166
76656-168
77623-196
2-(2-Propynyloxy)ethylamine 98% (by titrimetric analysis)
2-(2-Propynyloxy)ethylamine
Precaución: for Research Use Only
Formula:
C₅H₉NO |
MDL Number:
MFCD16713446 CAS Number: 122116-12-5 UN: 2734 ADR: 8,3,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...