tert-Butyl 4-(piperazine-1-carbonyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A961082-100MG A961082-1G A961082-250MG A961082-5G A961082-10G
76656-156EA 63.66 USD
76656-156 76656-158 76656-160 77599-308 77749-792
tert-Butyl 4-(piperazine-1-carbonyl)piperidine-1-carboxylate 98%
tert-Butyl 4-(piperazine-1-carbonyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇N₃O₃
MW: 297.39 g/mol
MDL Number: MFCD07367784
CAS Number: 887587-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR